Anti-Methyl Lysine Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-940 600-401-940
RL600-401-940EA 518.16 USD
CARL600401-940 RL600-401-940
Anti-Methyl Lysine Rabbit Polyclonal Antibody
Antibodies
This affinity-purified antibody is suitable for use in ELISA, western blotting and immunoprecipitation assays.  The antibody reacts specifically with methylated lysine residues.

Recommended Dilutions: ELISA: 1:1000 - 1:20000; Western Blot: 1:500 - 1:3000;

Type: Primary
Antigen: Methyl Lysine
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Avian, Mammalian
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR