Anti-MCM2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-922 600-401-922
RL600-401-922EA 518.16 USD
RL600-401-922 CA600-401-922
Anti-MCM2 Rabbit Polyclonal Antibody
Antibodies
This affinity purified antibody has been tested for use in ELISA and by western blot.  Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 100 kDa in size.

Recommended Dilutions: ELISA: 1:10000 - 1:50000; Western Blot: 1:500 - 1:2000;

Type: Primary
Antigen: MCM2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, S. cerevisiae
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR