Anti-MAPKAPK2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-281 600-401-281
RL600-401-281EA 518.16 USD
RL600-401-281 CA600-401-281
Anti-MAPKAPK2 Rabbit Polyclonal Antibody
Antibodies
This antibody is suitable for western blotting, ELISA, immunoprecipitation and kinase assays.

Recommended Dilutions: ELISA: 1:15000; Western Blot: 1:2000;

Type: Primary
Antigen: MAPKAPK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR