Anti-IRAK3 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-954 600-401-954
RL600-401-954EA 518.29 USD
RL600-401-954 CARL600401-954
Anti-IRAK3 Rabbit Polyclonal Antibody
Antibodies
Anti-IRAK-M Antibody is suitable for use in ELISA, immunochemistry and western blot.  Expect a band approximately 68 kDa in size corresponding to IRAK-M Protein by western blotting in the appropriate cell lysate or extract.

Recommended Dilutions: ELISA: 1:5000 - 1:20000; Western Blot: 1:500 - 1:2000; Immunochemistry: 1:300 - 1:1000;

Type: Primary
Antigen: IRAK3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR