Anti-IRAK2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-952 600-401-952
RL600-401-952EA 518.29 USD
RL600-401-952 CARL600401-952
Anti-IRAK2 Rabbit Polyclonal Antibody
Antibodies
This affinity-purified antibody has been tested for use in ELISA, immunochemistry and western blot.  Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 65 kDa in size corresponding to IRAK-2 Protein.

Recommended Dilutions: ELISA: 1:5000 - 1:20000; Western Blot: 1:500 - 1:1000; Immunochemistry: 1:300 - 1:600;

Type: Primary
Antigen: IRAK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR