Anti-IDN3 Goat Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-101-288 600-101-288
RL600-101-288EA 518.16 USD
CA600-101-288 RL600-101-288
Anti-IDN3 Goat Polyclonal Antibody
Antibodies
This affinity purified antibody has been tested for use in ELISA and western blot.  Specific conditions for reactivity should be optimized by the end user. Expect bands at 316 kDa in size corresponding to IDN3 by western blotting.

Recommended Dilutions: ELISA: 1:2000 - 1:10000; Western Blot: 1:500 - 1:3000;

Type: Primary
Antigen: IDN3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR