Anti-PCNA Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-968 600-401-968
RL600-401-968EA 518.16 USD
RL600-401-968 CA600-401-968
Anti-PCNA Rabbit Polyclonal Antibody
Antibodies
This affinity purified antibody has been tested for use in ELISA and western blot. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 29-36 kDa in size.

Recommended Dilutions: ELISA: 1:2000 - 1:8000; Western Blot: 1:500 - 1:2000;

Type: Primary
Antigen: PCNA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Chicken, Dog, Human, Monkey, Mouse, Rat, Xenopus, fish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR