Anti-ING1 Goat Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-101-282 600-101-282
RL600-101-282EA 518.16 USD
RL600-101-282 CA600-101-282
Anti-ING1 Goat Polyclonal Antibody
Antibodies
This affinity-purified antibody has been tested for use in ELISA and western blotting using over expressed cell lysates. Expect a band approximately 47 kDa, 33 kDa or 24 kDa in size corresponding to variants of ING1 by western blotting.

Recommended Dilutions: ELISA: 1:4000 - 1:14000; Western Blot: 1:500 - 1:3000;

Type: Primary
Antigen: ING1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR