Anti-NF2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-414 600-401-414
RL600-401-414EA 518.16 USD
RL600-401-414 CA600-401-414
Anti-NF2 Rabbit Polyclonal Antibody
Antibodies
This phospho-specific polyclonal antibody was tested by immunoblotting and ELISA.  By ELISA, the antibody was found to be reactive with the phosphorylated form of the immunizing peptide and minimally reactive with the non-phosphorylated form of the immuni

Recommended Dilutions: ELISA: 1:1200,000; Western Blot: 1:500- 1:2000;

Type: Primary
Antigen: NF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR