Anti-IgG Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
609-4118 609-4118
RL609-4118EA 220.11 USD
RL609-4118 CA609-4118
Anti-IgG Rabbit Polyclonal Antibody
Antibodies
Anti-Human IgG F(ab')2 antibody is suitable for western blotting, Immunochemistryand for ELISA. Researchers should determine optimal titers for applications that are not stated below.

Recommended Dilutions: ELISA: 1:350000; Western Blot: 1:3000 - 1:15000; Immunochemistry: 1:1000 - 1:5000;

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: F(ab')2
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR