Anti-IgG Goat Polyclonal Antibody

Supplier: Rockland Immunochemicals
609-1116 609-1116
RL-609-1116EA 161.52 USD
RL-609-1116 CA609-1116
Anti-IgG Goat Polyclonal Antibody
Antibodies
Secondary Goat Anti-IgG (H&L) Reacts with Human

Recommended Dilutions: ELISA: 1:40000 - 1:160000; Western Blot: 1:1000 - 1:5000; Immunochemistry: 1:1000 - 1:2000; Other Dilution: User Optimized

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR