Anti-GSK3A Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-444 600-401-444
RL600-401-444EA 518.16 USD
CA600-401-444 RL600-401-444
Anti-GSK3A Rabbit Polyclonal Antibody
Antibodies
This affinity-purified antibody has been tested for use in ELISA and by western blot.  Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 52 kDa in size corresponding to GSK3A by western blotting.

Recommended Dilutions: ELISA: 1:4000 - 1:20000; Western Blot: 1:500 - 1:2000;

Type: Primary
Antigen: GSK3A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR