Anti-FANCC Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-672 600-401-672
RL600-401-672EA 518.16 USD
CA600-401-672 RL600-401-672
Anti-FANCC Rabbit Polyclonal Antibody
Antibodies
This affinity purified antibody has been tested for use in ELISA and by western blot.  Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 63 kDa in size corresponding to FANCC by western blotting.

Recommended Dilutions: ELISA: 1:20000 - 1:80,000; Western Blot: 1:1000 - 1:3000;

Type: Primary
Antigen: FANCC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Chimpanzee, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR