Anti-ATAD5 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-688 600-401-688
RL600-401-688EA 518.16 USD
RL600-401-688 CARL600401-688
Anti-ATAD5 Rabbit Polyclonal Antibody
Antibodies
This affinity purified antibody has been tested for use in ELISA and western blot.  Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 120 kDa in size corresponding to Elg1 by western blotting.

Recommended Dilutions: ELISA: 1:3000 - 1:15000; Western Blot: 1:500 - 1:2000;

Type: Primary
Antigen: ATAD5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR