Anti-DNMT3L Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-454 600-401-454
RL600-401-454EA 518.16 USD
CA600-401-454 RL600-401-454
Anti-DNMT3L Rabbit Polyclonal Antibody
Antibodies
This affinity purified antibody has been tested for use in ELISA against the immunizing peptide.  Reactivity in other immunoassays is unknown.

Recommended Dilutions: ELISA: 1:1000 - 1:3000; Western Blot: 1:500; Immunochemistry: 1:500; Immunoprecipitation: 1:100;

Type: Primary
Antigen: DNMT3L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR