Anti-NCAPH Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-682 600-401-682
RL600-401-682EA 518.16 USD
RL600-401-682 CARL600401-682
Anti-NCAPH Rabbit Polyclonal Antibody
Antibodies
This affinity purified antibody has been tested for use in ELISA and by western blot.  Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 83 kDa in size corresponding to human BRRN1 by western blotting.

Recommended Dilutions: ELISA: 1:8000 - 1:35000; Western Blot: 1:500 - 1:2000;

Type: Primary
Antigen: NCAPH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR