Anti-AP1G1 Goat Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-101-289 600-101-289
RL600-101-289EA 518.16 USD
CA600-101-289 RL600-101-289
Anti-AP1G1 Goat Polyclonal Antibody
Antibodies
This affinity purified antibody has been tested for use in ELISA and by western blot.  Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 91 kDa in size corresponding to AP1G1 by western blotting.

Recommended Dilutions: ELISA: 1:2000 - 1:8000; Western Blot: 1:500 - 1:2000; Immunoprecipitation: 1:100;

Type: Primary
Antigen: AP1G1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR