Anti-IgG Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
608-4103 608-4103
RL608-4103EA 189.2 USD
CA608-4103 RL608-4103
Anti-IgG Rabbit Polyclonal Antibody
Antibodies
Secondary Rabbit Anti-IgG F(c) Reacts with Horse (Equine)

Recommended Dilutions: ELISA: 1:2000,000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: F(c)
Host: Rabbit
Isotype: IgG
Reactivity: Horse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR