Anti-HIF1A Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-898 600-401-898
RL600-401-898EA 518.16 USD
RL600-401-898 CA600-401-898
Anti-HIF1A Rabbit Polyclonal Antibody
Antibodies
This affinity purified antibody has been tested for use in ELISA, immunochemistry and by western blot.  Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 17 kDa in size.

Recommended Dilutions: ELISA: 1:5000 - 1:25000; Western Blot: 1:500 - 1:2000; Immunochemistry: 20 µg/ml;

Type: Primary
Antigen: HIF1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR