Anti-GSK3B Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-424 600-401-424
RL600-401-424EA 522.84 USD
RL600-401-424 CA600-401-424
Anti-GSK3B Rabbit Polyclonal Antibody
Antibodies
This antibody has been tested for use in ELISA, immunochemistry, western blotting and immunoprecipitation.  Reactivity in other immunoassays is unknown.  Serum starved 293T whole cell lysate is suitable for use as a positive control.

Recommended Dilutions: ELISA: 1:10000 - 1:50000; Immunofluorescence: 1:200 - 1:1000; Western Blot: 1:500 - 1:3000; Immunochemistry: 1:200 - 1:1000;

Type: Primary
Antigen: GSK3B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR