Anti-IgG Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
605-4102 605-4102
RL605-4102EA 154.28 USD
CA605-4102 RL605-4102
Anti-IgG Rabbit Polyclonal Antibody
Antibodies
Secondary antibody reagents are ideal for western blotting, immunochemistry, ELISA, Fluorescence Microscopy, Flow Cytometry as well as other antibody detection methods.

Recommended Dilutions: ELISA: 1:800,000; Western Blot: 1:10000 - 1:60,000; Immunochemistry: 1:1000 - 1:5000;

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy and Light chain
Host: Rabbit
Isotype: IgG
Reactivity: Goat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR