Anti-FANCF Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-674 600-401-674
RL600-401-674EA 518.16 USD
CARL600401-674 RL600-401-674
Anti-FANCF Rabbit Polyclonal Antibody
Antibodies
This affinity purified antibody has been tested for use in ELISA and by western blot.  Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 42 kDa in size corresponding to FANCF by western blotting.

Recommended Dilutions:

Type: Primary
Antigen: FANCF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Internal
Host: Rabbit
Isotype: IgG
Reactivity: Chimpanzee, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR