Anti-IgG Goat Polyclonal Antibody

Supplier: Rockland Immunochemicals
616-1102 616-1102
RL616-1102EA 154.28 USD
RL616-1102 CA616-1102
Anti-IgG Goat Polyclonal Antibody
Antibodies
Secondary Goat Anti-IgG (H&L) Reacts with Donkey

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Donkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR