Anti-CREB1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-270 600-401-270
RL600-401-270EA 518.16 USD
RL600-401-270 CA600-401-270
Anti-CREB1 Rabbit Polyclonal Antibody
Antibodies
Anti-CREB pS133 antibody reacts with phosphorylated human CREB at pS133 and shows minimal reactivity by western blot with non-phosphorylated CREB and minimal reactivity (1%) by ELISA against the non-phosphorylated form of the immunizing peptide.

Recommended Dilutions: ELISA: 1:1000 - 1:6000; Western Blot: 1:500 - 1:2000; Immunochemistry: 20 µg/ml;

Type: Primary
Antigen: CREB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR