Anti-IgG Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
601-4103 601-4103
RL601-4103EA 189.23 USD
RL601-4103 CA601-4103
Anti-IgG Rabbit Polyclonal Antibody
Antibodies
Bovine IgG F(c) Antibody is suitable for immunoblotting (western or dot blot), ELISA, and immunochemistry requiring extremely low background levels, lot-to-lot consistency, high titer and specificity.

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: F(c)
Host: Rabbit
Isotype: IgG
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR