Anti-CTCFL Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-907 600-401-907
RL600-401-907EA 518.16 USD
RL600-401-907 CA600-401-907
Anti-CTCFL Rabbit Polyclonal Antibody
Antibodies
BORIS affinity purified antibody has been tested for use in ELISA and by western blot.  Specific conditions for reactivity should be optimized by the end user. Expect a predominant band approximately 75 kDa in size corresponding to BORIS by western blotting.

Recommended Dilutions: ELISA: 1:2000 - 1:10000; Western Blot: 1:200 - 1:2000;

Type: Primary
Antigen: CTCFL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR