Anti-ASAP1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-910 600-401-910
RL600-401-910EA 518.16 USD
RL600-401-910 CA600-401-910
Anti-ASAP1 Rabbit Polyclonal Antibody
Antibodies
This affinity purified antibody has been tested for use in ELISA, immunochemistry, Immunofluorescence microscopy and by western blot.  Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 130 kDa in size.

Recommended Dilutions: ELISA: 1:4000 - 1:16000; Western Blot: 1:500 - 1:2000; Immunochemistry: 20-40 µg/ml;

Type: Primary
Antigen: ASAP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Chicken, Chimpanzee, Dog, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR