Anti-ASAP1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-911 600-401-911
RL600-401-911EA 518.16 USD
RL600-401-911 CA600-401-911
Anti-ASAP1 Rabbit Polyclonal Antibody
Antibodies
This affinity purified antibody has been tested for use in ELISA, Immunofluorescence microscopy and by western blot.  Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 130 kDa in size corresponding to ASAP1 Protein by western blotting.

Recommended Dilutions: ELISA: 1:2000 - 1:10000; Western Blot: 1:500 - 1:2000;

Type: Primary
Antigen: ASAP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Chicken, Chimpanzee, Dog, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR