Anti-APOC2 Goat Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-101-113 600-101-113
RL600-101-113EA 1351.09 USD
CA600-101-113 RL600-101-113
Anti-APOC2 Goat Polyclonal Antibody
Antibodies
Anti-apoLipoProtein antibodies have been used for indirect trapping ELISA for quantitation of antigen in serum using a standard curve, immunoprecipitation, immunochemistry, and for western blotting for highly sensitive qualitative analysis.

Recommended Dilutions: ELISA: 1:5000 - 1:10000; Western Blot: 1:5000 - 1:10000; Immunochemistry: 1:50 - 1:200, 5µg/ml; Immunoprecipitation: 1:100;

Type: Primary
Antigen: APOC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR