Anti-APOA1 Goat Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-101-109 600-101-109
RL600-101-109EA 1351.08 USD
CA600-101-109 RL600-101-109
Anti-APOA1 Goat Polyclonal Antibody
Antibodies
Anti-apoLipoProtein antibodies have been used for indirect trapping ELISA for quantitation of antigen in serum using a standard curve, for immunoprecipitation, immunochemistry and for western blotting for highly sensitive qualitative analysis.

Recommended Dilutions: ELISA: 1:10000 - 1:20000; Western Blot: 1:1000 - 1:2000; Immunochemistry: 1:50 - 1:200; Immunoprecipitation: 1:100;

Type: Primary
Antigen: APOA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR