Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-AP2A2 Goat Polyclonal Antibody

Supplier: Rockland Immunochemical
600-101-287
This affinity purified antibody has been tested for use in ELISA and by western blot.  Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 104-107 kDa in size corresponding to AP2A1 or AP2A2 Proteins by western blotting.

Recommended Dilutions: ELISA: 1:5000 - 1:20000; Western Blot: 1:500 - 1:2000; Immunoprecipitation: 1:100;

Type: Primary
Antigen: AP2A2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Chicken, Chimpanzee, Dog, Human, Mouse, Rat, orangutan




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 µg 600-101-287 RL600-101-287 Each (100µG) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol AP2A2
Antigen Name Adaptor-Related Protein Complex 2, Alpha 2 Subunit
Reactivity Human, Chimpanzee, Mouse, Rat, Chicken, Dog, Orangutan
Conjugation Unconjugated
Clonality Polyclonal
Antibody Type Primary
Host Goat
Isotype IgG
Western Blot Yes
ELISA Yes
ImmunoPrecipitation Yes
Antigen Synonyms Adapter-related protein complex 2 alpha-2 subunit, HYPJ, HIP9, Alpha-adaptin C, AP2A2, Alpha2-adaptin, CLAPA2, 100 kDa coated vesicle protein C, Adaptor protein complex AP-2 subunit alpha-2, ADTAB, AP-2 complex subunit alpha-2, Clathrin assembly, KIAA0899
Amino Acid Number 3 to 14
Gene ID 161
Concentration 1.7 mg/ml by UV absorbance at 280 nm
Format Antibody fraction
Cross Adsorption No
Immunogen This affinity purified antibody was prepared from whole goat serum produced by repeated immunizations with a synthetic peptide corresponding aa 3-14 of Human AP2A1 and AP2A2 Proteins.
Purification affinity-purified
Form Liquid (sterile filtered)
Storage Buffer 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide,
Storage Temperature -20°C
Shipping Temperature Dry Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty