Anti-AKT1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-269 600-401-269
RL600-401-269EA 491.65 USD
RL600-401-269 CA600-401-269
Anti-AKT1 Rabbit Polyclonal Antibody
Antibodies
AKT antibody is phospho specific for pT308 and is suitable for western blotting and ELISA.

Recommended Dilutions: ELISA: 1:15000;

Type: Primary
Antigen: AKT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR