Anti-ABCB6 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-945 600-401-945
RL600-401-945EA 518.16 USD
RL600-401-945 CARL600401-945
Anti-ABCB6 Rabbit Polyclonal Antibody
Antibodies
This affinity-purified antibody has been tested for use in ELISA and western blot.  Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 94 kDa in size corresponding to ABCB6 Protein by western blotting.

Recommended Dilutions: ELISA: 1:40000 - 1:130,000; Western Blot: 1:1000 - 1:5000;

Type: Primary
Antigen: ABCB6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Dog, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR