Anti-ABCB1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-967 600-401-967
RL600-401-967EA 518.16 USD
RL600-401-967 CA600-401-967
Anti-ABCB1 Rabbit Polyclonal Antibody
Antibodies
This affinity purified antibody has been tested for use in ELISA and western blot.

Recommended Dilutions: ELISA: 1:10000 - 1:50000; Western Blot: 1:200 - 1:2000;

Type: Primary
Antigen: ABCB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Dog, Human, Monkey, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR