Anti-MLLT4 Rabbit Polyclonal Antibody

Supplier: Abcepta
AP6191D
60942-652EA 751.67 USD
60942-652
Anti-MLLT4 Rabbit Polyclonal Antibody
Antibodies
Primary Rabbit Anti-Afadin (MLLT4) (Center A1152) Reacts with Human

Type: Primary
Antigen: MLLT4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center A1152
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR