Anti-AF4 Rabbit Polyclonal Antibody

Supplier: Abcepta
AP6189A
80089-022EA 789.25 USD
80089-022
Anti-AF4 Rabbit Polyclonal Antibody
Antibodies
Primary Rabbit Anti-AF4 (MLLT2) (C-term) Reacts with Human

Type: Primary
Antigen: AF4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR