VWR® Talon® Universal Stirrer Mounting Bracket
915511
60079-103CS 170.88 USD
60079-103
VWR® Talon® Universal Stirrer Mounting Bracket
This aluminum mounting bracket is used to mount stirrers and other devices weighing 2 kg (4.4 lbs.) or less to lab frames, ring stands, and more.
  • Aluminum construction
  • Versatile utility
  • Compatible with many mounting options
  • Maximum grip size 0 – 20 mm (0 – 25/32")
Aluminum mounting bracket with attachment screws.