Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VWR® Talon® Heavy-Duty Tapered Clamps

Supplier: VWR International
915291 915290
Clamps have four prongs, dual adjustments, and a tapered design to hold large vessels with ground-glass joint necks.

  • Clamps avoid wobbling and vibrations
  • Tapered design securely holds large vessels
  • Easy to attachment to lab frames
  • Nickel-plated zinc construction

Easy attachment to lab frames and other apparatus.

Accessories information: Thumbscrew knobs feature a 32 mm (11/4") slot in the center that will accept thumbscrews supplied with the VWR® Talon® multi-purpose clamps, excluding the stainless steel version. Thumbscrew knobs can be press-fit onto screws to create a small knob for clamping or securing, which makes tightening of clamps easier on the hands and wrists.

Ordering information: An additional holder must be purchased in order to attach clamps to frames or other apparatus.



 

ORDER

Expand All / Collapse All
Size Overall Length Maximum Grip Size VWR Catalog Number Unit Price Quantity
Large 381 mm (15") 34/45 28641-020 Each Retrieving
Product available on GSA Advantage®
Small 356 mm (14") 24/40 28641-018 Each Retrieving
Product available on GSA Advantage®
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Arm Ø 13 mm (1/2")
Arm Length 229 mm (9")

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty