    Sort by:
  • Search Within Results
You Searched for: "ethanol-absolute-denatured-with-4-6-isopropanol"
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
SearchResultCount:"1"
Sort by:
Supplier:  Alfa Aesar
Description:   Specially denatured alcohol (SDA) formula 3A consists of 5 volumes of methanol and 100 volumes of 200 proof ethanol.