    Sort by:
  • Search Within Results
Methylidenecyclopentane
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
Sort by:
Supplier:  TCI America
Description:   CAS Number: 1528-30-9 MDL Number: MFCD00001421 Molecular Formula: C6H10 Molecular Weight: 82.15 Purity/Analysis Method: 98.0% (GC) Form: Clea...