SevenCompact™ pH/Ion Meters, S220, METTLER TOLEDO®
Supplier: Mettler-Toledo
SevenCompact™
30019029 30019032 30019032 30019028 30019031 30019028 30019029 30019031
89207-238EA 1566.52 USD
CA89207-240 89207-238 89207-236 CA89207-234 89207-234 CA89207-236 CA89207-238 89207-240
SevenCompact™ pH/Ion Meters, S220, METTLER TOLEDO®
Electrochemical Meters pH and Multi-Parameter Meters
The SevenCompact™ series combines precise electrochemical measurement technologies with flexibility, innovative design and ease-of-use. Optimal measuring specifications ensure great performance in nearly all pH or conductivity related laboratory measurements.
  • Fast and accurate measurements
  • User-friendly operation
  • Menu guidance in 13 languages
  • Clear, well arranged color display
  • Many peripheral options
  • GLP compliant
  • Service offering, including IQ/OQ
The SevenCompact™ pH/Ion meter S220 is the perfect choice for a wide range of applications. It is much more than a pH meter: it can also measure ORP and ion concentration in various units. Starting a measurement or calibration has never been easier, requiring only a single keypress.
The optional robust and reliable sensor InLab Expert Pro-ISM is designed for the measurement of samples with the most diverse properties.
The optional refillable glass sensor InLab Routine Pro-ISM is the perfect choice for proteinaceous and other precious and demanding samples.
The optional low-maintenance InLab Versatile Pro is suited for customers that need an accurate yet economic sensor for routine measurements.
Ordering information: The meter-only model includes an electrode holder, protective cover, operating instructions, quick guide, declaration of conformity and test certificate. The kits also include sensor, guide to pH measurements and 2 buffer sachets for pH 4.01, 7.00, 9.21 and 10.00, in addition to the meter-only standard accessories.