Would you like to log in again?
Accessories
Supplier: Expedeon
Flexicast
WGT-1420 WGT-1412 WGT-1416 WGT-1408
ORDER
Expand All / Collapse All
Description No. of Slots Pathlength Supplier No. VWR Catalog Number Unit Price Quantity
Replacement Winged Gel Tray, 14 x 12 cm 3 3 cm WGT-1412 71002-976 Each Retrieving Restricted
Minority or Woman-Owned Business Enterprise Small Business Enterprise
Replacement Winged Gel Tray, 14 x 16 cm 4 3 cm WGT-1416 71002-978 Each Retrieving Restricted
Minority or Woman-Owned Business Enterprise Small Business Enterprise
Replacement Winged Gel Tray, 14 x 20 cm 6 2.6 cm WGT-1420 71002-980 Each Retrieving Restricted
Minority or Woman-Owned Business Enterprise Small Business Enterprise
Replacement Winged Gel Tray, 14 x 8 cm 2 2.6 cm WGT-1408 71002-974 Each Retrieving Restricted
Minority or Woman-Owned Business Enterprise Small Business Enterprise
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:
  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address
* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
ACCESSORIES
Expand All / Collapse All
Accessories
Description Details Supplier No. VWR Catalog Number Unit Price Quantity
Combs for 14 cm Unit 1.0 mm x 12-Well, (Thickness x No. of Wells) SGC14-1012 CBSGC14-1012 Each Retrieving Restricted
Combs for 14 cm Unit 1.0 mm x 16-Well, (Thickness x No. of Wells) SGC14-1016 CBSGC14-1016 Each Retrieving Restricted
Combs for 14 cm Unit 1.0 mm x 20-Well, (Thickness x No. of Wells) SGC14-1020 CBSGC14-1020 Each Retrieving Restricted
Combs for 14 cm Unit 1.0 mm x 26-Well, (Thickness x No. of Wells) SGC14-1026 CBSGC14-1026 Each Retrieving Restricted
Combs for 14 cm Unit 1.0 mm x 27-Well, (Thickness x No. of Wells) SGC14-1027 CBSGC14-1027 Each Retrieving Restricted
Combs for 14 cm Unit 1.5 mm x 10-Well, (Thickness x No. of Wells) SGC14-1510 CBSGC14-1510 Each Retrieving Restricted
Combs for 14 cm Unit 1.5 mm x 12-Well, (Thickness x No. of Wells) SGC14-1512 CBSGC14-1512 Each Retrieving Restricted
Combs for 14 cm Unit 1.5 mm x 16-Well, (Thickness x No. of Wells) SGC14-1516 CBSGC14-1516 Each Retrieving Restricted
Combs for 14 cm Unit 1.5 mm x 20-Well, (Thickness x No. of Wells) SGC14-1520 CBSGC14-1520 Each Retrieving Restricted
Combs for 14 cm Unit 1.5 mm x 26-Well, (Thickness x No. of Wells) SGC14-1526 CBSGC14-1526 Each Retrieving Restricted
Combs for 14 cm Unit 1.5 mm x 27-Well, (Thickness x No. of Wells) SGC14-1527 CBSGC14-1527 Each Retrieving Restricted
Combs for 14 cm Unit 2.0 mm x 10-Well, (Thickness x No. of Wells) SGC14-2010 CBSGC14-2010 Each Retrieving Restricted
Combs for 14 cm Unit 2.0 mm x 12-Well, (Thickness x No. of Wells) SGC14-2012 CBSGC14-2012 Each Retrieving Restricted
Combs for 14 cm Unit 2.0 mm x 20-Well, (Thickness x No. of Wells) SGC14-2020 CBSGC14-2020 Each Retrieving Restricted
Combs for 14 cm Unit 2.0 mm x 26-Well, (Thickness x No. of Wells) SGC14-2026 CBSGC14-2026 Each Retrieving Restricted
Combs for 14 cm Unit 2.0 mm x 27-Well, (Thickness x No. of Wells) SGC14-2027 CBSGC14-2027 Each Retrieving Restricted
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Q and A
Product Questions & Answers
Sort by: Most Helpful | Newest | Oldest
Items Per Page: 5  10  20 
2  3  4  Next
Can't find an answer to your question?  Ask us a question
Ask us a question:
Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question
Q&A Guidelines & Tips
If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.
When writing your question or answer, please follow these guidelines:
  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English
All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s
We reserve the right not to post your question if it contains any of the following
  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty