Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Opti-Pak, 1.0µL Trap Column, Optimize Technologies

Supplier: OPTIMIZE
10-03225-TA 10-03225-TB 10-03225-TG 10-03225-TC 10-03225-TD 10-03225-TE 10-03225-TF
The Opti-Pak is specially designed for use in today's low volume applications

The unique hardware design integrates a PEEK holder with an auto-adjusting stem which provides a fluid interface with any 10-32 standard or microinjector port, and guarantees a zero-dead-volume connection despite variances in tubestop depth.

Installing an Opti-Pak into the outlet of a switching valve will facilitate the trapping, desalting, and pre-concentration of an analyte mixture. With Opti-Pak's versatility, a second trap can be added downstream in the loop of an injection valve and effectively perform a 2D separation. The elegant, flexible design of Opti-Pak makes it amenable to virtually any trapping application. Opti-Pak can be installed directly into the fitting port of an injection valve. Two or more Opti-Paks can also be installed side by side in the same valve stator without interference.

With bed volumes ranging from 0.12 µL to 5.0 µL, you can find the perfect balance between sample capacity and trap volume. A proprietary method of frit placement allows the packed stem to be positioned directly into the flow path, which eliminates all excess swept volume. Less swept volume translates to absolute minimum sample dilution and dispersion.



 

ORDER

Expand All / Collapse All
Description Column Format Phase Capacity I.D. x L Flow Rate Packing Material Bed Weight Supplier No. VWR Catalog Number Unit Price Quantity
Opti-Pak Trap Column Analytical C8 1.0 µL 3.2 x 50 mm 10.0 - 40.0 µL/min Silica 4.0 µg 10-03225-TC 89209-612 Pack of 5 Retrieving
Opti-Pak Trap Column Analytical C18 1.0 µL 3.2 x 150 mm 10.0 - 40.0 µL/min Silica 4.0 µg 10-03225-TG 89209-638 Pack of 5 Retrieving
Opti-Pak Trap Column Analytical C18AQ 1.0 µL 2.1 x 50 mm 10.0 - 40.0 µL/min Silica 4.0 µg 10-03225-TB 89209-610 Pack of 5 Retrieving
Opti-Pak Trap Column Analytical C4 1.0 µL 4 x 50 mm 10.0 - 40.0 µL/min Silica 4.0 µg 10-03225-TD 89209-614 Pack of 5 Retrieving
Opti-Pak Trap Column Analytical SAX 1.0 µL 2.1 x 150 mm 10.0 - 40.0 µL/min Silica 4.0 µg 10-03225-TF 89209-618 Pack of 5 Retrieving
Opti-Pak Trap Column Analytical SCX 1.0 µL 4.6 x 50 mm 10.0 - 40.0 µL/min Silica 4.0 µg 10-03225-TE 89209-616 Pack of 5 Retrieving
Opti-Pak Trap Column Analytical C18 1.0 µL 4.6 x 100 mm 10.0 - 40.0 µL/min Silica 4.0 µg 10-03225-TA 89209-608 Pack of 5 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty