1-Heptadecanol

Supplier: Thermo Scientific Chemicals

Synonyms: Heptadecan-1-olHexadecyl carbinol

L20404-14 L20404-14 L20404-06 L20404-06
AAAL20404-14EA 214.99 USD
AAAL20404-14 CAAAAL20404-06 AAAL20404-06 CAAAAL20404-14
1-Heptadecanol
1-Heptadecanol
Formula: H₃C(CH₂)₁₅CH₂OH
MW: 256.47 g/mol
Boiling Pt: 308…309 °C
Melting Pt: 52…56 °C
Flash Pt: 155 °C(311 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002822
CAS Number: 1454-85-9
EINECS: 215-932-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR