2,3,5,6-Tetrafluoro-4-hydroxybenzoic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

L20209-14 L20209-14 L20209-03 L20209-06 L20209-06 L20209-03
AAAL20209-14EA 502.64 USD
AAAL20209-14 AAAL20209-03 CAAAAL20209-06 CAAAAL20209-14 CAAAAL20209-03 AAAL20209-06
2,3,5,6-Tetrafluoro-4-hydroxybenzoic acid 97%
2,3,5,6-Tetrafluoro-4-hydroxybenzoic acid
Formula: C₇H₂F₄O₃
MW: 210.08 g/mol
Melting Pt: 148…150 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00129952
CAS Number: 652-34-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR