2,4-Dibromo-1-fluorobenzene 98%

Warning

Synonyms: 1,3-Dibromo-4-fluorobenzene1-Fluoro-2,4-dibromobenzene

L20171-06 L20171-14 L20171-06 L20171-14
AAAL20171-06EA 107.19 USD
AAAL20171-06 CAAAAL20171-14 AAAL20171-14 CAAAAL20171-06
2,4-Dibromo-1-fluorobenzene 98%
2,4-Dibromo-1-fluorobenzene
Formula: C₆H₃Br₂F
MW: 253.9 g/mol
Boiling Pt: 103 °C/32 mm
Density: 2.047
Flash Pt: 92 °C(197 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000283
CAS Number: 1435-53-6
EINECS: 215-860-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR