3-Bromo-2,4,5-trifluorobenzoic acid 98%

Warning

L20154-03 L20154-14 L20154-03 L20154-14 L20154-06 L20154-06
AAAL20154-03EA 51.49 USD
AAAL20154-03 AAAL20154-14 CAAAAL20154-06 AAAL20154-06 CAAAAL20154-14 CAAAAL20154-03
3-Bromo-2,4,5-trifluorobenzoic acid 98%
3-Bromo-2,4,5-trifluorobenzoic acid
Formula: C₇H₂BrF₃O₂
MW: 254.99 g/mol
Melting Pt: 122…123 °C
Storage Temperature: Ambient
MDL Number: MFCD05664065
CAS Number: 104222-42-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR