2-Fluoro-3-pyridineboronic acid 97%

Warning

Synonyms: 2-Fluoro-3-pyridinylboronic acid(2-Fluoropyridin-3-yl)boronic acid2-Fluoropyridine-3-boronic acid

L20108-03 L20108-06 L20108-03 L20108-06
AAAL20108-03EA 48.19 USD
AAAL20108-03 CAAAAL20108-06 AAAL20108-06 CAAAAL20108-03
2-Fluoro-3-pyridineboronic acid 97%
2-Fluoro-3-pyridineboronic acid
Formula: C₅H₅BFNO₂
MW: 140.91 g/mol
Melting Pt: 172 °C
Storage Temperature: Ambient
MDL Number: MFCD03411557
CAS Number: 174669-73-9
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR