2-Fluoro-6-nitrotoluene 98%

Warning

L20093-22 L20093-22 L20093-06 L20093-06 L20093-14 L20093-14
AAAL20093-22EA 268.62 USD
AAAL20093-22 CAAAAL20093-14 AAAL20093-14 CAAAAL20093-22 CAAAAL20093-06 AAAL20093-06
2-Fluoro-6-nitrotoluene 98%
2-Fluoro-6-nitrotoluene
Formula: C₇H₆FNO₂
MW: 155.13 g/mol
Boiling Pt: 208…210 °C
Melting Pt: 6…8 °C
Density: 1.277
Flash Pt: 88 °C(190 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007197
CAS Number: 769-10-8
EINECS: 212-203-3
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR