2,3,6-Trifluoro-4-(trifluoromethyl)pyridine 97%

Supplier: Thermo Scientific Chemicals
Warning

L20053-03 L20053-MD L20053-MD L20053-03
AAAL20053-MDEA 105.53 USD
CAAAAL20053-MD AAAL20053-MD CAAAAL20053-03 AAAL20053-03
2,3,6-Trifluoro-4-(trifluoromethyl)pyridine 97%
2,3,6-Trifluoro-4-(trifluoromethyl)pyridine
Formula: C₆HF₆N
MW: 201.07 g/mol
Boiling Pt: 104…106 °C
Density: 1.579
Storage Temperature: Ambient
MDL Number: MFCD03840203
CAS Number: 84940-46-5
UN: 1993
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR